Skip to Content
Merck
2-Phenylpyridine 98%

2-Phenylpyridine

2-Phenylpyridine 98%
Empirical Formula (Hill Notation):
C11H9N
CAS Number:
Molecular Weight:
155.20
EC Number:

Products

Product Category
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing