Skip to Content
Merck
2-Nonanone natural, ≥97%, FCC, FG

2-Nonanone

2-Nonanone natural, ≥97%, FCC, FG
Synonym(s):
Heptyl methyl ketone
Linear Formula:
CH3(CH2)6COCH3
CAS Number:
Molecular Weight:
142.24
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing