
2-Nitrophenyl octyl ether

Synonym(s):
1-Nitro-2-octyloxybenzene
Linear Formula:
O2NC6H4O(CH2)7CH3
CAS Number:
Molecular Weight:
251.32
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Format
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing