
2-Methacryloyloxyethyl phosphorylcholine

Synonym(s):
MPC
Empirical Formula (Hill Notation):
C11H22NO6P
CAS Number:
Molecular Weight:
295.27
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing