Skip to Content
Merck
2-Iminothiolane hydrochloride ≥98% (TLC), powder

2-Iminothiolane hydrochloride

2-Iminothiolane hydrochloride ≥98% (TLC), powder
Synonym(s):
2-Thiolanimine hydrochloride, 2IT, Dihydro-2(3H)-thiophenimine hydrochloride, Traut’s reagent
Empirical Formula (Hill Notation):
C4H7NS · HCl
CAS Number:
Molecular Weight:
137.63

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing