Skip to Content
Merck
2-Hydroxymethyl-12-crown-4 95%

2-Hydroxymethyl-12-crown-4

2-Hydroxymethyl-12-crown-4 95%
Synonym(s):
(12-Crown-4)-2-methanol, 1,4,7,10-Tetraoxacyclododecan-2-methanol
Empirical Formula (Hill Notation):
C9H18O5
CAS Number:
Molecular Weight:
206.24
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing