
2-(Dodecylthiocarbonothioylthio)-2-methylpropionic acid

Synonym(s):
2-(Dodecylthiocarbonothioylthio)-2-methylpropanoic acid, S-Dodecyl-S′-(α,α′-dimethyl-α′′-acetic acid)trithiocarbonate, DDMAT
Empirical Formula (Hill Notation):
C17H32O2S3
CAS Number:
Molecular Weight:
364.63
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing