
2-Deoxy-D-glucose

Synonym(s):
2-Deoxy-D-arabinohexose, 2-Deoxy-D-glucose
Empirical Formula (Hill Notation):
C6H12O5
CAS Number:
Molecular Weight:
164.16
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Research Area
Quality Segment
Application
Optical Isomer
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing