2-Cyanoethyl N,N-diisopropylchlorophosphoramidite
Synonym(s):
Chloro(diisopropylamino)-β-cyanoethoxyphosphine
Linear Formula:
[(CH3)2CH]2NP(Cl)OCH2CH2CN
CAS Number:
Molecular Weight:
236.68
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Product Line
Chemical Composition
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing