
2-Chlorotrityl chloride, polymer-bound

Synonym(s):
2,α-Dichlorobenzhydryl-polystyrene crosslinked with divinylbenzene
CAS Number:
Products
Brand
Application
Markush Class
Markush Group
Cleavage Condition
Linker
Chain Type
Reaction Type
Showing 1-3 of 3
Compare
Product Number
Description
Pricing