Skip to Content
Merck
2-Chlorophenyl phosphorodichloridate 99%

2-Chlorophenyl phosphorodichloridate

2-Chlorophenyl phosphorodichloridate 99%
Synonym(s):
2-Chlorophenyl dichlorophosphate
Linear Formula:
Cl2P(O)OC6H4Cl
CAS Number:
Molecular Weight:
245.43
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing