Skip to Content
Merck
2-Bromopyrazine 97%

2-Bromopyrazine

2-Bromopyrazine 97%
Synonym(s):
Bromopyrazine
Empirical Formula (Hill Notation):
C4H3BrN2
CAS Number:
Molecular Weight:
158.98

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing