
2-Aminopentane

Synonym(s):
1-Methylbutylamine, 2-Amylamine
Linear Formula:
CH3CH(NH2)CH2CH2CH3
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing