
2-Acetamido-2-deoxy-β-D-glucopyranosylamine

Synonym(s):
2-Acetamido-1-amino-1,2-dideoxy-β-D-glucopyranose, N-Acetyl-D-glucosylamine
Empirical Formula (Hill Notation):
C8H16N2O5
CAS Number:
Molecular Weight:
220.22
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing