Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
2,6-Dichloroquinone-4-chloroimide
Synonym(s):
2,6-Dichloroquinone-4-chloroimide,
N
,2,6-Trichloro-
p
-benzoquinoneimide, Gibb’s reagent
Empirical Formula (Hill Notation):
C
6
H
2
Cl
3
NO
CAS Number:
101-38-2
Molecular Weight:
210.45
EC Number:
202-937-2
Products
Brand
Supelco (2)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Application
Special Grade
Puriss (1)
Puriss P.A. (1)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
1.03037
2,6-Dichloroquinone-4-chloroimide,
GR for analysis reagent for Vitamin B₆ Reag. Ph Eur
Expand
View Pricing
35620
2,6-Dichloroquinone-4-chloroimide,
for spectrophotometric det. of vitamin B
6
, ≥99.0%
Expand
View Pricing