2,5-Furandicarboxylic acid
Empirical Formula (Hill Notation):
C6H4O5
CAS Number:
Molecular Weight:
156.09
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing