Skip to Content
Merck
2,4,6-Trifluorobenzenesulfonyl chloride 97%

2,4,6-Trifluorobenzenesulfonyl chloride

2,4,6-Trifluorobenzenesulfonyl chloride 97%
Empirical Formula (Hill Notation):
C6H2ClF3O2S
CAS Number:
Molecular Weight:
230.59

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing