Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
2,3-Diaminonaphthalene
Synonym(s):
2,3-Naphthalenediamine, DAN
Empirical Formula (Hill Notation):
C
10
H
10
N
2
CAS Number:
771-97-1
Molecular Weight:
158.20
EC Number:
212-241-0
Products
Product Category
dyes (1)
fluorescence reagent (1)
Brand
Sigma-Aldrich (2)
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
88461
2,3-Diaminonaphthalene,
BioReagent, suitable for fluorescence, ≥98.0% (HPLC)
Expand
View Pricing
D2757
2,3-Diaminonaphthalene,
≥95% purity (HPLC), powder
Expand
View Pricing