
22(R)-Hydroxycholesterol

Synonym(s):
22α-Hydroxycholesterol, 5-Cholestene-3β,22(R)-diol, 5-Cholestene-3β,22[R]-diol
Empirical Formula (Hill Notation):
C27H46O2
CAS Number:
Molecular Weight:
402.65
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Purity
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing