
2,2-Dimethylpropylmagnesium chloride solution

Synonym(s):
Neopentylmagnesium chloride
Linear Formula:
(CH3)3CCH2MgCl
CAS Number:
Molecular Weight:
130.90
Products
Product Category
Formula Weight
Physical Form
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing