Skip to Content
Merck
2,2′-Azobis(2-methylpropane) 97%

2,2′-Azobis(2-methylpropane)

2,2′-Azobis(2-methylpropane) 97%
Synonym(s):
Azo-tert-butane, Di-tert-butyldiazene
Linear Formula:
(CH3)3CN=NC(CH3)3
CAS Number:
Molecular Weight:
142.24

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing