Skip to Content
Merck
(1S)-(+)-Ketopinic acid 99%

(1S)-(+)-Ketopinic acid

(1S)-(+)-Ketopinic acid 99%
Synonym(s):
(1S)-7,7-Dimethyl-2-oxobicyclo[2.2.1]heptane-1-carboxylic acid
Empirical Formula (Hill Notation):
C10H14O3
CAS Number:
Molecular Weight:
182.22

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing