Skip to Content
Merck
1-Docosanol 98%

1-Docosanol

1-Docosanol 98%
Synonym(s):
Behenyl alcohol, 1-Docosanol
Linear Formula:
CH3(CH2)21OH
CAS Number:
Molecular Weight:
326.60

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing