Skip to Content
Merck
1-Chloroisoquinoline 95%

1-Chloroisoquinoline

1-Chloroisoquinoline 95%
Empirical Formula (Hill Notation):
C9H6ClN
CAS Number:
Molecular Weight:
163.60

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing