
1-Butyl-3-methylimidazolium thiocyanate

Synonym(s):
1-Butyl-3-methyl-1H-imidazolium thiocyanate
Empirical Formula (Hill Notation):
C9H15N3S
CAS Number:
Molecular Weight:
197.30
Products
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing