1-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulfonyl)imide
Empirical Formula (Hill Notation):
C11H20F6N2O4S2
CAS Number:
Molecular Weight:
422.41
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing