Skip to Content
Merck
1-Bromo-2-methylpropane 99%

1-Bromo-2-methylpropane

1-Bromo-2-methylpropane 99%
Synonym(s):
Isobutyl bromide, 1-Bromo-2-methylpropane
Linear Formula:
(CH3)2CHCH2Br
CAS Number:
Molecular Weight:
137.02

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing