Skip to Content
Merck
1-Bromo-2,4,6-triisopropylbenzene 95%

1-Bromo-2,4,6-triisopropylbenzene

1-Bromo-2,4,6-triisopropylbenzene 95%
Synonym(s):
2,4,6-Triisopropylbromobenzene, 2-Bromo-1,3,5-triisopropylbenzene
Empirical Formula (Hill Notation):
C15H23Br
CAS Number:
Molecular Weight:
283.25

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing