Skip to Content
Merck
1,4-Dioxane solution suitable for NMR (reference standard), 5 mM in chloroform-d (99.8 atom % D), NMR tube size 3 mm × 8 in.

1,4-Dioxane solution

1,4-Dioxane solution suitable for NMR (reference standard), 5 mM in chloroform-d (99.8 atom % D), NMR tube size 3 mm × 8 in.
Synonym(s):
1,4-Dioxane solution
Empirical Formula (Hill Notation):
C4H8O2
CAS Number:
Molecular Weight:
88.11

Products

Product Category
Brand
Formula Weight
Physical Form
Format
Chemical Composition
Quality Segment
Markush Class
Markush Group
Special Grade
Technique
Showing 1-10 of 10
Compare
Product Number
Description
Pricing