
1,4-Dioxane-d8

Synonym(s):
p-Dioxane-d8, Octadeuterodioxane
Empirical Formula (Hill Notation):
C4D8O2
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Isotope
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing