Skip to Content
Merck
1,4-Dinitrobenzene 98%

1,4-Dinitrobenzene

1,4-Dinitrobenzene 98%
Synonym(s):
1,4-Dinitrobenzene
Linear Formula:
1,4-(NO2)2C6H4
CAS Number:
Molecular Weight:
168.11

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing