
1,4-Cyclohexanedicarboxylic acid

Linear Formula:
C6H10(CO2H)2
CAS Number:
Molecular Weight:
172.18
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing