Skip to Content
Merck
1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic acid ≥97.0% (CHN)

1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic acid

1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic acid ≥97.0% (CHN)
Synonym(s):
DOTA
Empirical Formula (Hill Notation):
C16H28N4O8 · xH2O
CAS Number:
Molecular Weight:
404.42 (anhydrous basis)

Products

Brand
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing