Skip to Content
Merck
1,2-Bis(3-aminopropylamino)ethane technical grade, 94%

1,2-Bis(3-aminopropylamino)ethane

1,2-Bis(3-aminopropylamino)ethane technical grade, 94%
Synonym(s):
3,3′-Ethylenediiminodipropylamine
Linear Formula:
NH2CH2CH2CH2NHCH2CH2NHCH2CH2CH2NH2
CAS Number:
Molecular Weight:
174.29
EC Number:

Products

Product Category
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing