
11-Aminoundecanoic acid

Synonym(s):
11-Aminoundecanoic acid
Linear Formula:
H2N(CH2)10COOH
CAS Number:
Molecular Weight:
201.31
Products
Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Amino Acid Type
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing