
1,1,3-Trichloroacetone

Synonym(s):
1,1,3-Trichloro-2-propanone
Linear Formula:
(Cl)2CHCOCH2Cl
CAS Number:
Molecular Weight:
161.41
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing