Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
10L Tedlar
®
PLV Gas Sampling Bag w/Thermogreen
®
LB-2 Septa
Synonym(s):
Tedlar
®
Gas Sampling Bag
Products
Product Category
bags (1)
Brand
Supelco (1)
Agency
Length (in)
Application
Technique
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
24634
10L Tedlar
®
PLV Gas Sampling Bag w/Thermogreen
®
LB-2 Septa,
maximum volume 10 L, Push/Pull Lock Valve (PLV) (Push Lock Valve (PLV))
Expand
View Pricing