Skip to Content
Merck
10,12-Tricosadiynoic acid ≥98.0% (GC)

10,12-Tricosadiynoic acid

10,12-Tricosadiynoic acid ≥98.0% (GC)
Empirical Formula (Hill Notation):
C23H38O2
CAS Number:
Molecular Weight:
346.55

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing