G006195
HPLC Analysis of Sugars on SUPELCOSIL™ LC-NH2
application for HPLC
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
technique(s)
HPLC: suitable
test parameters
column: SUPELCOSIL LC-NH2, 25 cm x 4.6 mm I.D., 5 μm particles (58338)
mobile phase: [A] water; [B] acetonitrile; (25:75, A:B)
flow rate: 1 mL/min
temp.: ambient
detector: refractive index (RI)
injection: 2 μL
sample: 10 mg/mL in water
suitability
application for HPLC
application(s)
food and beverages
Legal Information
SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC
Analyte
Description
Fructose, United States Pharmacopeia (USP) Reference Standard
Dextrose, United States Pharmacopeia (USP) Reference Standard
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%
Maltose solution, for molecular biology, BioReagent, ~20% in H2O
Lactose, pharmaceutical secondary standard; traceable to USP and PhEur
analytical column
Product No.
Description
Pricing
standard
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service