24535
Hamilton® syringe, 700 series, cemented needle, pt.#5
710N, volume 100 μL, needle size 22s ga, needle L 51 mm (2 in.)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122001
eCl@ss:
32070500
Recommended Products
material
cone tip stainless steel needle (point style 5)
needle size
22s ga
feature
needle type N (cemented needle)
point style 5
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 80639
needle L
51 mm (2 in.)
volume
100 μL
Looking for similar products? Visit Product Comparison Guide
General description
- Hamilton® syringe, 700 series, cemented needle, pt.#5 allows liquid samples to be filled and dispensed through the side hole.
- Septum damage is minimized by the solid domed tip.
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service