All Photos(1)
About This Item
Linear Formula:
[C6H10O5]n
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.24
Recommended Products
grade
analytical standard
for GPC
Quality Level
form
powder or crystals
mol wt
Mn ~18,300
Mp ~21,400
Mw ~23,800
analyte chemical class(es)
oligosaccharides
technique(s)
gel permeation chromatography (GPC): suitable
Mw/Mn
~1.30
application(s)
food and beverages
InChI
1S/C18H32O16/c19-1-5(21)9(23)10(24)6(22)3-31-17-16(30)14(28)12(26)8(34-17)4-32-18-15(29)13(27)11(25)7(2-20)33-18/h1,5-18,20-30H,2-4H2
InChI key
FZWBNHMXJMCXLU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Dextran is a polysaccharide. Dextran fluids behave as colloids; and also are used as plasma substitution. It may have siliconizing effect helping in coating raw serosal surfaces.
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
The Peritoneum
The Peritoneum, 324-324 (2012)
Yoshikazu Isomura et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(25), 10209-10220 (2013-06-21)
It is widely accepted that dorsal striatum neurons participate in either the direct pathway (expressing dopamine D1 receptors) or the indirect pathway (expressing D2 receptors), controlling voluntary movements in an antagonistically balancing manner. The D1- and D2-expressing neurons are activated
Kelli G Sharp et al.
Experimental neurology, 257, 186-204 (2014-04-22)
As part of the NIH "Facilities of Research Excellence-Spinal Cord Injury" project to support independent replication, we repeated key parts of a study reporting robust engraftment of neural stem cells (NSCs) treated with growth factors after complete spinal cord transection
Krystal Nizar et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(19), 8411-8422 (2013-05-10)
Calcium-dependent release of vasoactive gliotransmitters is widely assumed to trigger vasodilation associated with rapid increases in neuronal activity. Inconsistent with this hypothesis, intact stimulus-induced vasodilation was observed in inositol 1,4,5-triphosphate (IP3) type-2 receptor (R2) knock-out (KO) mice, in which the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service