Z743672
Laser Tough-Tags™ microcentrifuge tube labels, kiss cut with square corners
white, pkg of 3200 labels 20 sheets of 160 labels, L × W 0.94 in. × 0.50 in.
Synonym(s):
solvent resistant labels, temperature resistant labels
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
55121611
NACRES:
NB.35
Recommended Products
packaging
pkg of 3200 labels 20 sheets of 160 labels
manufacturer/tradename
Diversified Biotech TTSW-KK-20
L × W
0.94 in. × 0.50 in.
color
white
Looking for similar products? Visit Product Comparison Guide
General description
Laboratory labels formatted for laser printers. Tough-Tags™ are chemically inert laboratory labels that strongly adhere to microtubes, test tubes, storage boxes and microscope slides. They resist organic solvents and caustic agents, making them perfect for accurately identifying important samples stored in moderate freezer temperatures.
Legal Information
Tough-Tags is a trademark of Diversified Biotech
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service