Skip to Content
Merck
All Photos(1)

Key Documents

SML3555

Sigma-Aldrich

Pirmenol

≥98% (HPLC)

Synonym(s):

CL-845, cis-(±)-α-[3-(2,6-Dimethyl-1-piperidinyl)propyl]-α-phenyl-2-pyridinemethanol, rel-α-​[3-​[(2R,​6S)​-​2,​6-Ddimethyl-​1-​piperidinyl]​propyl]​-​α-​phenyl-​2-​pyridinemethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H30N2O
CAS Number:
Molecular Weight:
338.49
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

, White to v. dark grey

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC(C1=CC=CC=C1)(C2=NC=CC=C2)CCCN3[C@@H](C)CCC[C@H]3C

InChI

1S/C22H30N2O/c1-18-10-8-11-19(2)24(18)17-9-15-22(25,20-12-4-3-5-13-20)21-14-6-7-16-23-21/h3-7,12-14,16,18-19,25H,8-11,15,17H2,1-2H3/t18-,19+,22?

InChI key

APUDBKTWDCXQJA-QIDMFYOTSA-N

Biochem/physiol Actions

Pirmenol is an antiarrhythmic agent class Ia that exhibit long half-life and low toxicities. Pirmenol suppresses premature ventricular contractions in human studies.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pirmenol: an antiarrhythmic drug with unique electrocardiographic features--a double-blind placebo-controlled comparison with quinidine
Salerno DM, Fifield J, Farmer C, Hodges M.
Clinical Cardiology (1991)
CL-845 (pirmenol hydrochloride): a new orally effective long-acting antiarrhythmic agent
Steffe TJ, Mertz TE, Hastings SG, Potoczak RE, Kaplan HR.
Journal of Pharmacology and Experimental Therapeutics (1980)
Pirmenol inhibits muscarinic acetylcholine receptor-operated K+ current in the guinea pig heart.
Watanabe Y, Hara Y, Tamagawa M, Nakaya H.
European Journal of Pharmacology (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service