M7150
Murashige and Skoog Vitamin Powder
1000 ×, powder, suitable for plant cell culture
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352207
NACRES:
NA.72
Recommended Products
form
powder
Quality Level
concentration
1000 ×
technique(s)
cell culture | plant: suitable
pH
3.5-4.5 (1.031 g/10 mL)
application(s)
agriculture
shipped in
ambient
storage temp.
2-8°C
Application
Murashige and Skoog medium is a widely used plant tissue culture growth medium. M&S Basal Medium contains macronutrients that include high levels of nitrate and organic additives such as agar, sugars, vitamins and growth regulators. Important growth regulators frequently added to M&S include IAA (auxin/morphogen) and the Kinetin (cytokinin/cell division promoter).
Preparation Note
Murashige and Skoog medium can be reconstituted from powder or by combining products that are major components of complete M&S medium, such as macronutrient mixtures and vitamin mixtures. Murashige and Skoog Vitamin powder (M7150) can be combined with Murashige and Skoog micro/macro-nutrients to generate basal media which when supplemented with organics, agar, auxins and cytokinins yield complete medium for growth plant tissue culture.
Reconstitution
Use at a concentration of one mL per liter of prepared medium to achieve the proper final concentration.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Elsa Matthus et al.
Plant physiology, 179(4), 1754-1767 (2019-01-31)
Phosphate (Pi) deficiency strongly limits plant growth, and plant roots foraging the soil for nutrients need to adapt to optimize Pi uptake. Ca2+ is known to signal in root development and adaptation but has to be tightly controlled, as it
Elizabeth J Carino et al.
Journal of virology, 94(22) (2020-08-28)
Maize chlorotic mottle virus (MCMV) combines with a potyvirus in maize lethal necrosis disease (MLND), a serious emerging disease worldwide. To inform resistance strategies, we characterized the translation initiation mechanism of MCMV. We report that MCMV RNA contains a cap-independent
Saxena et al.
Plant science : an international journal of experimental plant biology, 155(2), 133-140 (2000-05-18)
Efficient protocols have been established for both direct and indirect regeneration of plants in Pelargonium graveolens Indian cultivar Hemanti (Algerian type). Murashige and Skoog's (MS) medium [T. Murashige, F. Skoog, A revised medium for rapid growth and bioassays with tobacco
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service