GE27-1860-04
2′-Deoxycytidine 5′-Triphosphate, sodium (dCTP)
Cytiva 27-1860-04, pkg of 250 mg
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106305
NACRES:
NA.52
Recommended Products
form
solid
packaging
pkg of 250 mg
manufacturer/tradename
Cytiva 27-1860-04
shipped in
dry ice
storage temp.
−20°C
Related Categories
General description
2′-Deoxycytidine 5′-Triphosphate, sodium (dCTP).
Application
- For applications requiring dNTPs of high purity such as amplification, dideoxy sequencing, labeling, mutagenesis, cDNA synthesis, and expression profiling.
- High Purity dNTPs are functionally tested in long PCR* to be PCR-qualified, and meet or exceed criteria for high-quality sequencing with Thermo Sequenase DNA Polymerase.
- High purity: > 98% triphosphate, as confirmed by chromatographic analysis and 31P nuclear magnetic resonance (NMR) spectroscopy.
- These dNTPs are supplied as solids. For high purity solutions dNTPs, see the improved "illustra™ Solutions dNTPs".
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Analysis Note
To view the Certificate of Analysis for this product, please visit www.cytiva.com.
Legal Information
illustra is a trademark of Cytiva
Storage Class Code
13 - Non Combustible Solids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service