EP0030073398
Eppendorf® epT.I.P.S.® G Reloads, Eppendorf Quality™
volume (0.1-20 μL), L (40 mm), medium gray, pack of 960 ea (10 trays × 96 tips)
Synonym(s):
pipetting tips
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121600
NACRES:
NB.02
Recommended Products
material
gray (Tray)
sterility
non-sterile
packaging
pack of 960 ea (10 trays × 96 tips)
manufacturer/tradename
Eppendorf® 0030073398
L
(40 mm)
maximum volume
20 μL
volume
(0.1-20 μL)
color
medium gray
Looking for similar products? Visit Product Comparison Guide
General description
The pipette tips are single-use pipette tips for aspirating and dispensing liquids with air-cushion pipettors. The packaging of the pipette tips is color-coded and corresponds to the color of the control button on the Eppendorf pipettor.
Features and Benefits
- Minimized waste in comparison to disposable racks.
- The refill trays are autoclavable up to 121 °C, even when in packaging and without foil, allowing for reuse in subsequent applications.
- Depending on the tip size, the refill system is packaged either as dual-sided or in stack form, providing flexibility to meet different needs.
- Available in purity grades of Eppendorf® Quality and PCR clean.
- The trays are color-coded, simplifying volume and pipette identification for easy and efficient handling.
- Single-use pipette tips
- Packaged as reloade
- Universal cone
- Easy handling
- Avaiable in different quality grades
- Minimal force required for attachment or ejection
- Different volume sizes with color coding
- Volume marking for visual inspection
- Seal completely
- Provide maximum precision and accuracy
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Eppendorf Quality is a trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service