CLS6798420D
Corning® hotplate and stirrer with digital display
AC/DC input 230 V AC (CEE7-7 plug), plate L × W 5 in. × 7 in.
Synonym(s):
magnetic stirrer, magnetic stirrers, stirrers
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.72
Recommended Products
sterility
non-sterile
packaging
case of 1
manufacturer/tradename
Corning 6798-420D
availability
not available in USA, Canada and EU
parameter
5-550 °C temp. range
60-1150 rpm speed
plate L × W
5 in. × 7 in.
color
none
Looking for similar products? Visit Product Comparison Guide
General description
For use in Asia, Africa and Australia.
- Digital LED temperature display is adjustable in 5 °C increments and blinks until set temperature is reached
- Microprocessor maintains consistent and repeatable temperature and stir speed settings.
- Bright LED HOT TOP icon lights up when top plate temperature is over 60 °C, even when heat control is turned off.
- Separate temperature sensor provides power cut off if unit overheats
- Extremely durable and heat resistant Pyroceram glass-ceramic top
- Small footprint and low profile
- Meets UL and cUL standards
Legal Information
Corning is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service