C5791
Closures for 25 mm culture tubes, disposable
natural
Synonym(s):
closure cap, closure caps
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
Recommended Products
material
polypropylene
description
Autoclavable
color
natural
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Closures for 25 mm culture tubes, disposable made of polypropylene.
- Natural color.
- Available as a pack of 100 and 500 units.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service