Skip to Content
Merck
All Photos(2)

Key Documents

C1671

Sigma-Aldrich

Chlorprothixene hydrochloride

Synonym(s):

2-Chloro-9-(3-dimethylaminopropylidene)thioxanthene hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18ClNS · HCl
CAS Number:
Molecular Weight:
352.32
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

powder

Quality Level

storage temp.

2-8°C

SMILES string

Cl.CN(C)CC\C=C1\c2ccccc2Sc3ccc(Cl)cc13

InChI

1S/C18H18ClNS.ClH/c1-20(2)11-5-7-14-15-6-3-4-8-17(15)21-18-10-9-13(19)12-16(14)18;/h3-4,6-10,12H,5,11H2,1-2H3;1H/b14-7-;

InChI key

YWKRLOSRDGPEJR-KIUKIJHYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Chlorprothixene is a neuroleptic drug, which belongs to thioxanthene group. It has anticholinergic effects. Chlorprothixene might be associated with obstructive jaundice and parkinsonism. It is used to treat psychosis.

Application

Chlorprothixene hydrochloride has been used to study its exposure effects on learning and memory of rats.

Biochem/physiol Actions

D2 dopamine receptor antagonist; blocks a subset of GABAA receptors in rat cortex that is also blocked by clozapine; thioxanthine antipsychotic.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joshua B Melander et al.
Cell reports, 37(6), 109972-109972 (2021-11-11)
Cortical function relies on the balanced activation of excitatory and inhibitory neurons. However, little is known about the organization and dynamics of shaft excitatory synapses onto cortical inhibitory interneurons. Here, we use the excitatory postsynaptic marker PSD-95, fluorescently labeled at
Manavu Tohmi et al.
iScience, 26(1), 105778-105778 (2023-01-04)
Despite its importance, the development of higher visual areas (HVAs) at the cellular resolution remains largely unknown. Here, we conducted 2-photon calcium imaging of mouse HVAs lateromedial (LM) and anterolateral (AL) and V1 to observe developmental changes in visual response
M Froimowitz et al.
Journal of medicinal chemistry, 36(15), 2219-2227 (1993-07-23)
Conformational analyses have been performed on several phenothiazine and thioxanthene dopamine antagonists using the MM2-87 program and parameter set. The compounds that were examined are thioridazine (2), methotrimeprazine (3), cis- and trans-chlorprothixene, and a piperidylidene derivative of chlorprothixene. In addition
V Hadjimitova et al.
Pharmacology & toxicology, 84(4), 170-173 (1999-05-05)
The effect of some psychotropic drugs on the activity of macrophages to produce superoxide radicals during phagocytosis was tested. Three-cyclic antidepressants, imipramine and amitriptyline, and the thioxanthene neuroleptic, chlorprothixene, were studied. The superoxide production was measured by luminol-dependent chemiluminescence. The
Desk Reference of Clinical Pharmacology, Second Edition (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service