Skip to Content
Merck
All Photos(1)

Key Documents

PHL80356

Astilbin

phyproof® Reference Substance

Synonym(s):

Dihydroquercetin 3-rhamnoside, Taxifolin 3-rhamnoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22O11
CAS Number:
Molecular Weight:
450.39
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

manufacturer/tradename

PhytoLab

format

neat

storage temp.

2-8°C

SMILES string

O=C1C2=C(O)C=C(O)C=C2O[C@H](C3=CC(O)=C(O)C=C3)[C@H]1O[C@@]4([H])[C@H](O)[C@H](O)[C@@H](O)[C@H](C)O4

InChI

1S/C21H22O11/c1-7-15(26)17(28)18(29)21(30-7)32-20-16(27)14-12(25)5-9(22)6-13(14)31-19(20)8-2-3-10(23)11(24)4-8/h2-7,15,17-26,28-29H,1H3/t7-,15-,17+,18+,19-,20+,21-/m0/s1

InChI key

ZROGCCBNZBKLEL-MFSALPCASA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Biochem/physiol Actions

Flavonoid phytochemical found in St. John′s wort and Traditional Chinese Medicine herbal preparations. Immunosuppresive. Antiproliferative.

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qianni Duan et al.
Journal of ethnopharmacology, 249, 112405-112405 (2019-11-20)
The Chinese Herbal Prescription JieZe-1(JZ-1), added and subtracted from Yihuang Decoction, a famous formula in the 12th year of Kangxi in Qing Dynasty, has a clear effect on Genital Herpes (GH) and no obvious adverse reactions occur clinically. JZ-1 also
Meiyun Shi et al.
The Journal of pharmacy and pharmacology, 72(8), 1061-1071 (2020-04-30)
The purpose of this study is to reveal the pharmacokinetic profiles of astilbin with various doses in rats and investigate the oral absolute bioavailability and tissue distribution of astilbin after oral administration. Wistar rats were orally administered astilbin 12, 24 mg/kg
Yu Li et al.
International immunopharmacology, 84, 106571-106571 (2020-05-16)
Ischaemic stroke is a lethal cerebrovascular disease that occurs worldwide. Astilbin is a natural flavonoid compound with various physiological activities. The purpose of this study was to investigate the neuroprotective effects of Astilbin after cerebral ischaemia reperfusion (I/R) injury. The
Qingling Xie et al.
Journal of AOAC International, 103(1), 117-123 (2019-08-28)
Bupi Yiqi No. 1 Recipe (BPYQ), a traditional Chinese medicine formula, has been widely used as a treatment of multidrug resistance during chemotherapy in colorectal cancer and for the prevention of gastrointestinal cancer recurrence for decades. In this study, a
Dan Zheng et al.
Molecules (Basel, Switzerland), 25(20) (2020-10-21)
Astilbin and neoastilbin are two flavonoid stereoisomers. In the present study, their solubility, stability, and bioavailability were compared in a rat. The results revealed that the water solubility of astilbin and neoastilbin was 132.72 μg/mL and 217.16 μg/mL, respectively. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service